trans,trans-4'-Ethyl-4-(3,4,5-trifluorophenyl)bicyclohexyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:139215-80-8
:
Formula:C₂₀H₂₇F₃
MDL Number:MFCD12911825
:324.43 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:71.0 to 75.0 deg-C
- Mesomorphic range:71.0 to 77.0 deg-C
Specification Test Results
Frequently Bought Together