
Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate ≥98.0% (by GC)
: TCI
:155377-19-8
:
:C₇H₇F₃N₂O₂
:MFCD00052083
:208.14 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:143.0 to 147.0 deg-C