Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride ≥98.0% (by HPLC, total nitrogen)
: TCI
:4644-61-5
:
:C₈H₁₃NO₃
:MFCD00043267
:207.65 g/mol
:169°C
:225-073-8
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %