Order Entry
Finland
ContactUsLinkComponent
 
 
Ethyl-3-amino-1H-pyrazole-4-carboxylate ≥98.0% (by GC, titration analysis)
 :  TCI
 
 
Ethyl-3-amino-1H-pyrazole-4-carboxylate ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    101.0 to 106.0 deg-C