Ethyl-6-methyl-2-oxo-3-cyclohexene-1-carboxylate (mixture of isomers) ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3419-32-7
:
Formula:C₁₀H₁₄O₃
:182.22 g/mol
Density:1.07
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 95.0 %