Order Entry
Finland
ContactUsLinkComponent
 
 
1-Ethyl-4-methylcyclohexane (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC)
  TCIAE0424-1ML
 :  TCI
 :  
 
1-Ethyl-4-methylcyclohexane (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC)
  TCIAE0424-1ML
 :  TCI
 :  

 

  • Pk:
    1 mL
  • Seq:
    2070

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 % (total of isomers)

 

 

 :
 :126.24 g/mol
 :151°C
 :3295
 :3,III
 :GHS02
 :3728-56-1
 :C₉H₁₈
 :MFCD00045511