Order Entry
Finland
ContactUsLinkComponent
 
Flammable
1-Ethyl-3-methylcyclohexane (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC)
Catalog # TCIAE0423-1ML
Supplier:  TCI
CAS Number:  
Flammable
1-Ethyl-3-methylcyclohexane (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC)
Catalog # TCIAE0423-1ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 % (total of isomers)

Specification Test Results

About this item

 :
 :126.24 g/mol
Boiling Pt:150°C
UN:3295
ADR:3,III
 :GHS02
CAS Number:3728-55-0
Formula:C₉H₁₈
MDL Number:MFCD00045510