Ellagic acid dihydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:133039-73-3
:476-66-4
Formula:C₁₄H₆O₈
MDL Number:MFCD00006914
:302.19 g/mol
Merck Index:14,03547
EINECS:207-508-3
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Absorbance:min. 0.900 (5mg/L, ETOH, 254.0 to 258.0 nm) (calcd.on anh.substance)
- Absorbance2:min. 0.180 (5mg/L, ETOH, 365.0 to 369.0 nm) (calcd.on anh.substance)