Order Entry
Finland
ContactUsLinkComponent
 
 
1-Ethyl-1-cyclopentene ≥95.0% (by GC)
  TCIAE0293-5ML
 :  TCI
 :  
 
1-Ethyl-1-cyclopentene ≥95.0% (by GC)
  TCIAE0293-5ML
 :  TCI
 :  

 

  • Pk:
    5 mL
  • Seq:
    2210

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

 

 

 :
 :96.17 g/mol
 :107°C
 :3295
 :3,II
 :GHS02
 :2146-38-5
 :C₇H₁₂
 :MFCD00039453