
Di-tert-butyl phosphonate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13086-84-5
:
Formula:C₈H₁₉O₃P
MDL Number:MFCD00995019
Boiling Pt:66°C
Flash Pt:163°C
:194.21 g/mol
EINECS:235-996-8
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (Neutralization titration):min. 98.0 %
- Purity (Neutralization back titration):min. 98.0 %
Specification Test Results
Frequently Bought Together