
1,5-Dimethyl-1H-pyrazole-3-carboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5744-59-2
:
Formula:C₆H₈N₂O₂
MDL Number:MFCD00085040
:140.14 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:175.0 to 179.0 deg-C