Order Entry
Finland
ContactUsLinkComponent
10-[4-[4,6-Di(1-adamantyl)-1,3,5-triazin-2-yl]phenyl]-10H-spiro[acridine-9,9'-fluorene] ≥99.0% (by HPLC), purified by sublimation
10-[4-[4,6-Di(1-adamantyl)-1,3,5-triazin-2-yl]phenyl]-10H-spiro[acridine-9,9'-fluorene] ≥99.0% (by HPLC), purified by sublimation
Catalog # TCIAD5747-200MG
Supplier:  TCI
CAS Number:  
10-[4-[4,6-Di(1-adamantyl)-1,3,5-triazin-2-yl]phenyl]-10H-spiro[acridine-9,9'-fluorene] ≥99.0% (by HPLC), purified by sublimation
Catalog # TCIAD5747-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (HPLC):
    min. 99.0 area%

Specification Test Results

About this item

 :
 :755.02 g/mol
CAS Number:2250187-16-5
Formula:C₅₄H₅₀N₄