
1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
: TCI
:1036991-40-8
:
:C₁₁H₁₉BN₂O₂
:MFCD09864189
:222.0918 g/mol
:GHS07
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:85.0 to 89.0 deg-C