
Diallyl maleate ≥97.0% (by GC) stabilised
: TCI
:999-21-3
:
:C₁₀H₁₂O₄
:MFCD00010763
:196.21 g/mol
:154°C
:–47°C
:130°C
:213-658-0
:6.1,III
:Stabilised with hydroquinone (HQ)
:2810
:GHS06
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %