Order Entry
Finland
ContactUsLinkComponent
 
Dextran sulphate sodium salt, M.W. ~ 8000 (from dextran)
  TCIAD5143-25G
 :  TCI
 :  
Dextran sulphate sodium salt, M.W. ~ 8000 (from dextran)
  TCIAD5143-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Light yellow powder to crystal
  • Sulfur:
    15.0 to 20.0% (calcd.on dried substance)
  • Specific rotation [a]20/D:
    +75 to +105 deg (C=5, H2O,calcd.on dried substance)
  • limiting viscosity:
    0.018 to 0.032
  • Drying loss:
    max. 10.0 %

 

 

 :
 :14,02951
 :9011-18-1
 :(C₆H₇Na₃O₁₄S₃)n
 :MFCD00081551