3,5-Xylyl isothiocyanate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:40046-30-8
:
Formula:C₉H₉NS
MDL Number:MFCD00041082
:163.24 g/mol
UN:2810
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 97.0 %