trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:118164-51-5
:
Formula:C₂₃H₃₄F₂
MDL Number:MFCD13182310
:348.522 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:46.0 to 50.0 deg-C