trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:82832-57-3
:
Formula:C₂₁H₃₀F₂
MDL Number:MFCD09838997
:320.468 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:42.0 to 46.0 deg-C