Order Entry
Finland
ContactUsLinkComponent
4-(4,5-Diphenyl-1H-imidazol-2-yl)benzonitrile ≥98.0% (by HPLC, titration analysis)
Irritant
4-(4,5-Diphenyl-1H-imidazol-2-yl)benzonitrile ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD4655-200MG
Supplier:  TCI
CAS Number:  
Irritant
4-(4,5-Diphenyl-1H-imidazol-2-yl)benzonitrile ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD4655-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :321.38 g/mol
 :GHS07
CAS Number:29898-72-4
Formula:C₂₂H₁₅N₃