
Diethyl (trichloromethyl)phosphonate ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:866-23-9
:
Formula:C₅H₁₀Cl₃O₃P
MDL Number:MFCD00013666
:255.46 g/mol
Boiling Pt:135°C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together