
Diethylene Glycol Mono(2-propyn-1-yl) ether ≥97.0% (by GC)
Catalog # TCIAD4580-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
About this item
:
:144.17 g/mol
Boiling Pt:81°C
EINECS:230-612-5
:GHS07
CAS Number:7218-43-1
Formula:C₇H₁₂O₃
MDL Number:MFCD00053353