3,5-Dichlorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
: TCI
:68716-51-8
:
:C₁₂H₁₅BCl₂O₂
:MFCD05863906
:272.97 g/mol
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:51.0 to 55.0 deg-C