Order Entry
Finland
ContactUsLinkComponent
 
 
 
4-(Dimethylamino)butyronitrile ≥98.0% (by GC, titration analysis)
  TCIAD4546-1ML
 :  TCI
 :  
 
 
4-(Dimethylamino)butyronitrile ≥98.0% (by GC, titration analysis)
  TCIAD4546-1ML
 :  TCI
 :  

 

  • Pk:
    1 mL
  • Seq:
    2070

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

 

 

 :
 :112.18 g/mol
 :80°C
 :237-788-2
 :1993
 :3,III
 :GHS02
 :GHS07
 :13989-82-7
 :C₆H₁₂N₂
 :MFCD00019892