Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:59279-60-6
:
Formula:C₁₂H₂₁NO₆
MDL Number:MFCD03094774
:275.30 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:43.0 to 47.0 deg-C
- Specific rotation [a]20/D:+13.0 to +16.0 deg (C=1, CHCl3)