3,4-Difluoro-4'-(trans-4-propylcyclohexyl)biphenyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:85312-59-0
:
Formula:C₂₁H₂₄F₂
MDL Number:MFCD09838999
:314.42 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Mesomorphic range:68.0 to 100.0 deg-C