
3,3-Difluoropyrrolidine hydrochloride ≥98.0% (by titrimetric analysis)
: TCI
:163457-23-6
:
:C₄H₇F₂N
:MFCD03788948
:143.56 g/mol
:GHS07
- Appearance:White to Gray powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:132.0 to 136.0 deg-C
Frequently Bought Together