4-(4,5-Diphenyl-1H-imidazol-2-yl)phenol ≥98.0% (by HPLC, titration analysis) for biochemical research
Catalog # TCIAD4178-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Chemiluminescence test (Luminol/POD/H2O2):to pass test
Specification Test Results
About this item
:
:312.37 g/mol
CAS Number:1752-94-9
Formula:C₂₁H₁₆N₂O
MDL Number:MFCD00740542