Order Entry
Finland
ContactUsLinkComponent
2,5-Dihydro-4-methyl-2,5-dioxo-3-furanpropanoic acid ≥97.0% (by GC, titration analysis)
Irritant
2,5-Dihydro-4-methyl-2,5-dioxo-3-furanpropanoic acid ≥97.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
2,5-Dihydro-4-methyl-2,5-dioxo-3-furanpropanoic acid ≥97.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Methanolysis Method):
    min. 97.0 %
  • Melting point:
    97.0 to 101.0 deg-C

Specification Test Results

Related Information