Skip to Header
Skip to Main Content
Skip to Footer
Tarkennettu
Tarkennettu
SDS
Sertifikaatit
Valmistajan tiedot
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Kotisivu
2,7-Dibromo-9,9-di-n-octylfluorene
2,7-Dibromo-9,9-di-n-octylfluorene ≥98.0% (by HPLC)
2,7-Dibromo-9,9-di-n-octylfluorene ≥98.0% (by HPLC)
:
TCI
2,7-Dibromo-9,9-di-n-octylfluorene ≥98.0% (by HPLC)
:
TCI
:
198964-46-4
:
:
C₂₉H₄₀Br₂
:
MFCD03427216
:
548.45 g/mol
:
GHS07
Appearance:
White to Almost white powder to crystal
Purity (HPLC):
min. 98.0 area%
Melting point:
52.0 to 57.0 deg-C
Manual Synthetic Organic Chemistry and Materials Chemistry