
meso-1,2-Diphenylethylenediamine ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:951-87-1
:
Formula:C₁₄H₁₆N₂
MDL Number:MFCD00274328
:212.30 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:116.0 to 120.0 deg-C