
2,7-Dibromo-9,9-dimethylfluorene ≥98.0% (by HPLC)
: TCI
:28320-32-3
:
:C₁₅H₁₂Br₂
:MFCD00099471
:352.07 g/mol
:GHS07
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:178.0 to 182.0 deg-C
Frequently Bought Together