trans-4-(3,4-Difluorophenyl)-trans-4'-ethylbicyclohexane ≥97.0% (by GC)
: TCI
:118164-50-4
:MFCD13188635
:C₂₀H₂₈F₂
:MFCD11053389
:306.44 g/mol
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:51.0 to 54.0 deg-C