Order Entry
Finland
ContactUsLinkComponent
(1S,2S)-N,N'-Dimethyl-N,N'-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine ≥94.0% (by GC)
Irritant
(1S,2S)-N,N'-Dimethyl-N,N'-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine ≥94.0% (by GC)
Catalog # TCIAD3809-1G
Supplier:  TCI
CAS Number:  
Irritant
(1S,2S)-N,N'-Dimethyl-N,N'-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine ≥94.0% (by GC)
Catalog # TCIAD3809-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 94.0 %
  • Specific rotation [a]20/D:
    +29.0 to +35.0 deg (C=1, CHCl3)

Specification Test Results

About this item

 :
 :310.57 g/mol
Boiling Pt:145°C
 :GHS07
CAS Number:767291-67-8
Formula:C₂₀H₄₂N₂
MDL Number:MFCD11869717