1,4-Di(2-thienyl)-1,4-butanedione ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:13669-05-1
:
Formula:C₁₂H₁₀O₂S₂
MDL Number:MFCD00208362
:250.33 g/mol
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:132.0 to 137.0 deg-C