Order Entry
Finland
ContactUsLinkComponent
cis-Decahydro-1H,6H-3a,5a,8a,10a-tetraazapyrene ≥98.0% (by GC)
Irritant
cis-Decahydro-1H,6H-3a,5a,8a,10a-tetraazapyrene ≥98.0% (by GC)
Catalog # TCIAD3561-100MG
Supplier:  TCI
CAS Number:  
Irritant
cis-Decahydro-1H,6H-3a,5a,8a,10a-tetraazapyrene ≥98.0% (by GC)
Catalog # TCIAD3561-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    80.0 to 84.0 deg-C

Specification Test Results

About this item

 :
 :222.34 g/mol
 :GHS07
CAS Number:74199-16-9
Formula:C₁₂H₂₂N₄
MDL Number:MFCD04038934