Order Entry
Finland
ContactUsLinkComponent
 
1,1'-(Decane-1,10-diyl)bis[4-aza-1-azoniabicyclo[2.2.2]octane]dibromide ≥95.0% (by titrimetric analysis)
  TCIAD3269-1G
 :  TCI
 :  
1,1'-(Decane-1,10-diyl)bis[4-aza-1-azoniabicyclo[2.2.2]octane]dibromide ≥95.0% (by titrimetric analysis)
  TCIAD3269-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 95.0 %

 

 

 :
 :524.43 g/mol
 :94630-53-2
 :C₂₂H₄₄Br₂N₄