
(R)-1,2-Diaminopropane dihydrochloride ≥97.0% (by total nitrogen basis)
Catalog # TCIAD2827-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 97.0 %
- Specific rotation [a]20/D:+7.0 to +10.0 deg (C=1, MeOH)
Specification Test Results
About this item
:
:147.04 g/mol
:GHS07
CAS Number:19777-67-4
Formula:C₃H₁₀N₂
MDL Number:MFCD00239481