Order Entry
Finland
ContactUsLinkComponent
(S,S)-N,N'-Bis(trifluoromethanesulphonyl)-1,2-diphenylethylenediamine
Irritant
(S,S)-N,N'-Bis(trifluoromethanesulphonyl)-1,2-diphenylethylenediamine
Catalog # TCIAD2521-1G
Supplier:  TCI
CAS Number:  
Irritant
(S,S)-N,N'-Bis(trifluoromethanesulphonyl)-1,2-diphenylethylenediamine
Catalog # TCIAD2521-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :476.41 g/mol
 :GHS07
CAS Number:121788-77-0
Formula:C₁₆H₁₄F₆N₂O₄S₂
MDL Number:MFCD01321159