1,3-Dimethyl-2-thiohydantoin ≥98.0% (by GC)
Catalog # TCIAD2506-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:93.0 to 95.0 deg-C
Specification Test Results
About this item
:
:144.19 g/mol
CAS Number:1801-62-3
Formula:C₅H₈N₂OS
MDL Number:MFCD00186242