
(2-Butoxyethoxy)acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:82941-26-2
:
Formula:C₈H₁₆O₄
MDL Number:MFCD01105178
:176.21 g/mol
Boiling Pt:141°C
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %