
4-Diazo-4'-methoxydiphenylamine sulphate ≥98.0% (by HPLC)
Catalog # TCIAD2290-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
About this item
:
:323.32 g/mol
:GHS07
CAS Number:49732-38-9
Formula:C₁₃H₁₂N₃O
MDL Number:MFCD00080910