1-Dodecylhydantoin ≥98.0% (by GC)
Catalog # TCIAD2052-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:98.0 to 101.0 deg-C
Specification Test Results
About this item
:
:268.40 g/mol
EINECS:286-782-6
CAS Number:85391-28-2
Formula:C₁₅H₂₈N₂O₂
MDL Number:MFCD00142675