
1,3-Diphenyltetramethyldisilazane ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3449-26-1
:
Formula:C₁₆H₂₃NSi₂
MDL Number:MFCD00042916
:285.54 g/mol
Boiling Pt:146°C
Flash Pt:162°C
EINECS:222-372-5
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %