Order Entry
Finland
ContactUsLinkComponent
 
 
3,4-Dimethyl-1-pentyn-3-ol ≥96.0% (by GC)
  TCIAD1276-10ML
 :  TCI
 :  
undefined
 
3,4-Dimethyl-1-pentyn-3-ol ≥96.0% (by GC)
  TCIAD1276-10ML
 :  TCI
 :  

 

  • Pk:
    10 mL
  • Seq:
    2275

 

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 96.0 %

 

 

 :
 :112.17 g/mol
 :133°C
 :1987
 :3,III
 :GHS02
 :1482-15-1
 :C₇H₁₂O
 :MFCD00039845