3-(4-Chlorobenzal)phthalide ≥98.0% (by GC)
Catalog # TCIAC3448-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:153.0 to 158.0 deg-C
Specification Test Results
About this item
:
:256.69 g/mol
EINECS:243-862-5
CAS Number:20526-97-0
Formula:C₁₅H₉ClO₂
MDL Number:MFCD23115631