
trans-Cinnamic acid ≥99.5% (by GC, titration analysis) matrix for MALDI-TOF/MS
Catalog # TCIAC3412-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.5 %
- Purity (Neutralization titration):min. 99.5 %
- Melting point:132.0 to 136.0 deg-C
Specification Test Results
About this item
:
Merck Index:14,02299
Boiling Pt:300°C
:148.16 g/mol
EINECS:205-398-1
:GHS07
CAS Number:140-10-3
:621-82-9
Formula:C₉H₈O₂
MDL Number:MFCD00004369