
4-Chloro-5'-fluoro-2'-hydroxybenzophenone ≥97.0% (by GC)
: TCI
:62433-26-5
:
:C₁₃H₈ClFO₂
:MFCD00040948
:250.6500 g/mol
:263-539-2
:GHS07
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:66.0 to 70.0 deg-C