

4-Chloro-3-fluoropyridine ≥95.0% (by GC)
: TCI
:2546-56-7
:
:C₅H₃ClFN
:MFCD03453233
:131.54 g/mol
:139°C
:–24°C
:28°C
:Refrigerator
:1993
:3,III
:GHS02
:GHS07
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %