
4-(Cyanomethyl)phenylboronic acid (contains varying amounts of Anhydride)
: TCI
:91983-26-5
:
:C₈H₈BNO₂
:MFCD01632200
:160.97 g/mol
:191°C
:GHS07
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):98.0 to 113.0 %
Frequently Bought Together