Order Entry
Finland
ContactUsLinkComponent
2-Chloro-4-nitrobenzonitrile ≥98.0% (by GC)
Irritant
2-Chloro-4-nitrobenzonitrile ≥98.0% (by GC)
Supplier:  TCI
Certificates
Irritant
2-Chloro-4-nitrobenzonitrile ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    79.0 to 83.0 deg-C

Specification Test Results